missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TMEM199 Polyclonal antibody specifically detects TMEM199 in Human samples. It is validated for Western Blot, Immunofluorescence
Specifications
Specifications
| Antigen | TMEM199 |
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 532 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | C17orf32, chromosome 17 open reading frame 32, MGC45714, transmembrane protein 199 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human TMEM199 fragment. |
| Purification Method | Antigen and protein A Affinity-purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?