missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMEM194A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553157
This item is not returnable.
View return policy
Description
Immunogen sequence: LCTKNLEHPI QWLYITCRKV CKGAEKPVPP RLLTEEEYRI QGEVETRKAL EELREFCNSP DCSAWKTVSR IQSPKRFADF VEGSSHLTPN EVSVHEQEYG LGSIIAQDEI YEEASSEEED SYSRCPAITQ NNF Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 80%.
The function of this protein remains unknown.
Specifications
| TMEM194A | |
| Polyclonal | |
| Unconjugated | |
| NEMP1 | |
| Kiaa0286; NEMP1; Nuclear envelope integral membrane protein 1; Tmem194; Tmem194a; transmembrane protein 194; transmembrane protein 194A | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23306 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| O14524 | |
| NEMP1 | |
| Recombinant protein corresponding to Human TMEM194A. Recombinant protein control fragment (Product #RP-91173). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction