missing translation for 'onlineSavingsMsg'
Learn More

TMEM192 Antibody (S02-3F9), Novus Biologicals™

Rabbit Monoclonal Antibody

£328.00 - £500.00

Specifications

Antigen TMEM192
Clone S02-3F9
Concentration 0.3 mg/mL
Applications Western Blot, ELISA, Inhibition Assays
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18686123
View Documents
Novus Biologicals
NBP3-19787-100ul
100 μg
£500.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18658103
View Documents
Novus Biologicals
NBP3-19787-50ul
50 μg
£328.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

TMEM192 Monoclonal antibody specifically detects TMEM192 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, RNA Inhibition
Specifications

Specifications

TMEM192
0.3 mg/mL
Monoclonal
Purified
RUO
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
201931
IgG
Affinity purified
S02-3F9
Western Blot, ELISA, Inhibition Assays
Unconjugated
Rabbit
Human, Mouse, Rat
FLJ38482, transmembrane protein 192
Recombinant protein of human TMEM192 (Uniprot # Q8IY95)
Primary
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.