missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMEM185A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA561703
This item is not returnable.
View return policy
Description
Immunogen sequence: VGTGVWARNP QYRAEGETCV EFK Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
The protein encoded by this gene is predicted to be a transmembrane protein. This gene is best known for localizing to the CpG island of the fragile site FRAXF. The 5' untranslated region of this gene contains a CGG trinucleotide repeat sequence that normally consists of 7-40 tandem CGG repeats but which can expand to greater than 300 repeats. Methylation of the CpG island leads to transcriptional silencing of this gene, but neither the silencing nor an expanded repeat region appear to manifest itself in a clear phenotypic manner. Alternative splicing results in multiple transcript variants. A pseudogene of this gene has been defined on the X chromosome. [provided by RefSeq, Aug 2013].
Specifications
| TMEM185A | |
| Polyclonal | |
| Unconjugated | |
| TMEM185A | |
| BC023829; cDNA sequence BC023829; CXorf13; Ee3; erythropoietin-induced EST 3; FAM11A; family with sequence similarity 11, member A; fragile site, folic acid type, rare, fra(X)(q28) F; FRAXF; Protein FAM11A; Tmem185a; transmembrane protein 185A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 84548 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8NFB2 | |
| TMEM185A | |
| Recombinant protein corresponding to Human TMEM185A. Recombinant protein control fragment (Product #RP-100167). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction