missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TMEM184B Polyclonal Antibody
GREENER_CHOICE

Product Code. 12819082 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12819082 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12819082 Supplier Invitrogen™ Supplier No. PA520932

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is rat lung tissue lysate. PA5-20932 can be used with blocking peptide PEP-1046.

TMEM184B, also known as C22orf5, is a 407 amino acid multi-pass membrane protein and represents a novel gene in the activation of the MAPK signaling pathway. The gene encoding TMEM184B maps to human chromosome 22; mutations in several of the genes in chromosome 22 are involved in the development of autism, schizophrenia, Phelan-McDermid syndrome and Neurofibromatosis type 2, suggesting that TMEM184B may play a role in these syndromes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM184B
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene TMEM184B
Gene Accession No. Q8BG09, Q9Y519
Gene Alias 2610507A11; 4732495E13Rik; C22orf5; CTA-447C4.2; FM08; HS5O6A; HSPC256; PSEC0108; Putative MAPK-activating protein FM08; RGD1306591; Tmem184b; Transmembrane protein 184B
Gene Symbols TMEM184B
Host Species Rabbit
Immunogen A 15 amino acid peptide near the carboxy terminus of human TMEM184B.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 223693, 25829, 362959
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.