missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMEM16K Polyclonal Antibody

Description
Peptide sequence: LKADIDATLY EQVILEKEMG TYLGTFDDYL ELFLQFGYVS LFSCVYPLAA Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 92%.
TMEM16K is a multi-pass membrane protein. It belongs to the anoctamin family. TMEM16K may act as a calcium-activated chloride channel.
Specifications
Specifications
| Antigen | TMEM16K |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | Ano10 |
| Gene Accession No. | Q9NW15 |
| Gene Alias | AI604832; ANO10; ano10a; anoctamin 10; anoctamin 10a; anoctamin-10; fc37b04; hypothetical protein LOC566425; RGD1308260; SCAR10; Tmem16k; Transmembrane protein 16K; wu:fc37b04; zgc:114140 |
| Gene Symbols | Ano10 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?