missing translation for 'onlineSavingsMsg'
Learn More

TMEM16K Antibody (2B12-1A11), Novus Biologicals™

Product Code. 18369779 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18369779 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18369779 Supplier Novus Biologicals Supplier No. H00055129M01

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TMEM16K Monoclonal antibody specifically detects TMEM16K in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM16K
Applications Western Blot, ELISA
Classification Monoclonal
Clone 2B12-1A11
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA
Formulation In 1x PBS, pH 7.4
Gene Alias anoctamin 10, FLJ10375, MGC47890, Transmembrane protein 16KTMEM16Kanoctamin-10
Host Species Mouse
Immunogen FLJ10375 (AAH38855, 1 a.a. ~ 392 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MLLGAEAVGLVKECNDNTMRAFTYRTRQNFKGFDDNNDDFLTMAECQFIIKHELENLRAKDEKMIPGYPQAKLYPGKSLLRRLLTSGIVIQVFPLHDSEALKKLEDTWYTRFALKYQPIENHRLESAYQNHLILKVLVFNFLNCFASLFYIAFVLKDMKLLRQSLATLLITSQILNQIMESFLPYWLQRKHGVRVKRKVQALKADIDATLYEQVILEKEMGTYLGTFDDYLELFLQFGYVSLFSCVYPLAAAFAVLNNFTEVNSDALKMCRVFKRPFSEPSANIGVWQLAFEMMSVISVVTNCALIGMSPQVNAAFPESKADLILIVVAVEHALLALKFILAFAIPDKPRHIQMKLARLEFESLEALKQQQMKLVTENLKEEPMESGKEKAT
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55129
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.