Learn More
TMEM165 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA558331
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: REGLKMSPDE GQEELEEVQA ELKKKDEEFQ RTKLLNGPGD VETGTSITVP Q Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 92%.
The function of TM165 remains unknown.Specifications
TMEM165 | |
Polyclonal | |
Unconjugated | |
TMEM165 | |
AV026557; CDG2K; FT27; GDT1; pFT27; pt27; tm165; Tmem165; TMPT27; TPA regulated locus; Tpardl; TPA-regulated locus protein; Tparl; transmembrane protein 164; transmembrane protein 165; Transmembrane protein PFT27; transmembrane protein PT27; Transmembrane protein TPARL | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
55858 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9HC07 | |
TMEM165 | |
Recombinant protein corresponding to Human TMEM165 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only