missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMEM161B Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569919
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.
TMEM161B is a protein coding gene.
Specifications
| TMEM161B | |
| Polyclonal | |
| Unconjugated | |
| TMEM161B | |
| 2810446P07Rik; AI843389; FLB3342; id:ibd2207; PRO1313; RGD1309660; si:dkey-195m1.1; tmem161b; Transmembrane protein 161B; UNQ679/PRO1313; wu:fc31d04; zgc:63626 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 153396 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q8NDZ6 | |
| TMEM161B | |
| synthetic peptide directed towards the N terminal region of human TMEM161B. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction