missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMEM150C Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA571269
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 77%.
This gene encodes a transmembrane protein component of a mechanosensitve ion channel that is activated by mechanical stimuli in various cell types and confers slowly adapting, mechanically activated currents in dorsal root ganglion neurons. Mechanically activated ion channels are sensors that are critical for hearing, touch, pain, and blood pressure regulation.
Specifications
| TMEM150C | |
| Polyclonal | |
| Unconjugated | |
| TMEM150C | |
| gb:dn898596; RGD1306105; si:ch211-249h16.6; Tentonin 3; TMEM150C; Transmembrane protein 150C; Ttn3 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 441027 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| B9EJG8 | |
| TMEM150C | |
| synthetic peptide directed towards the C-terminal region of human TMEM150C. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction