missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMEM145 Polyclonal Antibody

Description
Antibody detects endogenous levels of total TMEM145.
TMEM145 gene ontology annotations related to this gene include G protein-coupled receptor signaling pathway; response to pheromone.
Specifications
Specifications
| Antigen | TMEM145 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | TMEM145 |
| Gene Accession No. | Q8C4U2, Q8NBT3 |
| Gene Alias | B930076A02Rik; Tmem145; Transmembrane protein 145 |
| Gene Symbols | TMEM145 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?