missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM132E Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-49639-25ul
This item is not returnable.
View return policy
Description
TMEM132E Polyclonal antibody specifically detects TMEM132E in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| TMEM132E | |
| Polyclonal | |
| Immunohistochemistry 1:500 - 1:1000, Immunohistochemistry-Paraffin 1:500 - 1:1000 | |
| DFNB99, Transmembrane Protein 132E | |
| This antibody was developed against a recombinant protein corresponding to amino acids: SHTILATTAAQQTLSFLKQEALLSLWLSYSDGTTAPLSLYSPRDYGLLVSSLDEHVATVTQDRAFPLVVAEAEGSGELL | |
| 25 μL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 124842 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction