missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TMEM132A Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA585903

Product Code. 16302815

  • £440.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

TMEM132A Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow

This gene encodes a protein that is highly similar to the rat Grp78-binding protein (GBP). Alternatively spliced transcript variants encoding different isoforms have been described.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

TMEM132A
Polyclonal
Unconjugated
TMEM132A
6720481D13Rik; GBP; glucose-regulated protein, 78kDa; GRP78 binding protein; GRP78-binding protein; heat shock 70kDa protein 5 (glucose-regulated protein, 78kDa) binding protein 1; heat shock 70kDa protein 5 binding protein 1; heat shock protein 5 binding protein 1; HSPA5-binding protein 1; Hspa5bp1; KIAA1583; LOW QUALITY PROTEIN: transmembrane protein 132A; Orai1; R74613; TMEM132A; Transmembrane protein 132A
Rabbit
Antigen affinity chromatography
RUO
54972, 98170
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
1 mg/mL
PBS with 0.2% BSA, 40% Glycerol and 0.05% sodium azide; pH 7.4
Q24JP5, Q922P8
TMEM132A
Synthetic peptide within Human Transmembrane protein 132A aa 204-253.
100 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.