missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMEM132A Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562524
This item is not returnable.
View return policy
Description
Immunogen sequence: QPVMGISLTL SRGTAHPGEV TATCWAQSAL PAPKQEVALS LWLSFSDHTV APAELYDRRD LGLSVSAEEP GAILPAEEQG AQLGVVVSGA GAEGLPLHVA L Highest antigen sequence identity to the following orthologs: Mouse - 92%, Rat - 90%.
This gene encodes a protein that is highly similar to the rat Grp78-binding protein (GBP). Alternatively spliced transcript variants encoding different isoforms have been described.
Specifications
| TMEM132A | |
| Polyclonal | |
| Unconjugated | |
| TMEM132A | |
| 6720481D13Rik; GBP; glucose-regulated protein, 78kDa; GRP78 binding protein; GRP78-binding protein; heat shock 70kDa protein 5 (glucose-regulated protein, 78kDa) binding protein 1; heat shock 70kDa protein 5 binding protein 1; heat shock protein 5 binding protein 1; HSPA5-binding protein 1; Hspa5bp1; KIAA1583; LOW QUALITY PROTEIN: transmembrane protein 132A; Orai1; R74613; TMEM132A; Transmembrane protein 132A | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 54972 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.20 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q24JP5 | |
| TMEM132A | |
| Recombinant protein corresponding to Human TMEM132A. Recombinant protein control fragment (Product #RP-100905). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction