Learn More
Invitrogen™ Tmem119 Monoclonal Antibody (V3RT1GOsz), Alexa Fluor™ 488, eBioscience™, Invitrogen™

Rat Monoclonal Antibody
£172.00 - £400.00
Specifications
| Antigen | Tmem119 |
|---|---|
| Clone | V3RT1GOsz |
| Concentration | 0.5 mg/mL |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17173983
|
Invitrogen™
53-6119-80 |
25 μg |
£172.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17183983
![]() |
Invitrogen™
53-6119-82 |
100 μg |
£400.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Description: This V3RT1GOsz monoclonal antibody recognizes mouse Tmem119. This clone has been reported to be successfully used in flow cytometry. Applications Reported: This V3RT1GOsz antibody has been reported for use in flow cytometric analysis. Applications Tested: This V3RT1GOsz antibody has been tested by flow cytometric analysis of mouse brain cells after myelin removal. This may be used at less than or equal to 1.0 μg per test. A test is defined as the amount (μg) of antibody that will stain a cell sample in a final volume of 100 μL. Cell number should be determined empirically but can range from 10^5 to 10^8 cells/test. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Excitation: 488 nm; Emission: 519 nm; Laser: Blue Laser
Tmem119 encodes a single-pass type I transmembrane protein which seems to interact with a multitude of micro-RNAs as well as certain transcription factors such as SMAD1, SMAD5 and RUNX2. Tmem119 has been identified as a surface marker of microglia that can be used to reliably distinguish microglia from infiltrating macrophages. Microglial cells are phagocytes that are restricted to the brain and involved in clearing up debris and dendrite pruning of neurons. They display a high functional plasticity, balancing neuroprotective and neurotoxic functions, which is reflected by their distinct physiology in response to changes in their microenvironment. Microglia are particularly mobile in areas of the brain undergoing restructuring. Reactive microglia co-express the marker Iba1. In a pathological context, microglia have been associated with differentiation of pathogenic/inflammatory A1 type astrocytes by secretion of Cq1, IL-1a and TNF when stimulated with LPS, and thought to contribute to the pathology of several neurodegenerative diseases such as Alzheimers, Parkinsons, Chorea Huntingtons disease and Multiple Sclerosis. The specific function Tmem119 plays on microglial cells remains unclear. Outside of the neural system, Tmem119 seems to be involved in osteoblast differentiation/ossification.Specifications
| Tmem119 | |
| 0.5 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 0.09% sodium azide; pH 7.2 | |
| AW208946; BC025600; OBIF; Osteoblast induction factor; PSEC0199; Tmem119; Transmembrane protein 119; UNQ731/PRO1415 | |
| TMEM119 | |
| IgG2a κ | |
| Affinity chromatography | |
| Antibody |
| V3RT1GOsz | |
| Flow Cytometry | |
| Alexa Fluor 488 | |
| Rat | |
| Mouse | |
| Q8R138 | |
| 231633 | |
| cell surface peptide. | |
| Primary | |
| 4°C, store in dark, DO NOT FREEZE! | |
| TMEM119 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.