missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMEM118 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA543500
This item is not returnable.
View return policy
Description
Peptide sequence: HGGHRGGSLL QHVGGDHRGH SEEGGDEQPG TPAPALSELK AVICWLQKGL Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 79%.
TMEM118 is a multi-pass membrane protein, and contains 1 RING-type zinc finger. The function of TMEM118 remains unknown.
Specifications
| TMEM118 | |
| Polyclonal | |
| Unconjugated | |
| RNFT2 | |
| AW049082; B830028P19Rik; hypothetical protein LOC553554; RGD1560195; RING finger and transmembrane domain-containing protein 2; RING finger and transmembrane domain-containing protein 2; ring finger protein, transmembrane 2; ring finger protein, transmembrane 2; RNFT2; si:dkey-175n1.1; Tmem118; Transmembrane protein 118; zgc:109947 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 84900 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q96EX2 | |
| RNFT2 | |
| Synthetic peptide directed towards the N-terminal of human TMEM118 (aa 118-167). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction