missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMEM11 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562483
This item is not returnable.
View return policy
Description
Immunogen sequence: ISWQFDPCCK YQVEYDAYKL SRLPLHTLTS STPVVLVRKD DLHRKRLHNT IAL Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
Plays a role in mitochondrial morphogenesis.
Specifications
| TMEM11 | |
| Polyclonal | |
| Unconjugated | |
| TMEM11 | |
| 5730466P16Rik; AA409091; C17orf35; PM1; PMI; Protein PM1; Protein PMI; putative receptor protein; RGD1309817; TMEM11; transmembrane protein 11; transmembrane protein 11, mitochondrial | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 8834 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P17152 | |
| TMEM11 | |
| Recombinant protein corresponding to Human TMEM11. Recombinant protein control fragment (Product #RP-101522). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction