missing translation for 'onlineSavingsMsg'
Learn More

TMEM11 Antibody, Novus Biologicals™

Product Code. 18330599 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
50 μg
Unit Size:
50µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18330599 50 μg 50µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18330599 Supplier Novus Biologicals Supplier No. H00008834B01P

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Polyclonal Antibody

TMEM11 Polyclonal antibody specifically detects TMEM11 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEM11
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS (pH 7.4)
Gene Accession No. NP_003867.1
Gene Alias C17orf35, chromosome 17 open reading frame 35, PM1putative receptor protein, PMI, Protein PM1, Protein PMI, transmembrane protein 11
Host Species Mouse
Immunogen TMEM11 (NP_003867.1, 1 a.a. - 192 a.a.) full-length human protein. MAAWGRRRLGPGSSGGSARERVSLSATDCYIVHEIYNGENAQDQFEYELEQALEAQYKYIVIEPTRIGDETARWITVGNCLHKTAVLAGTACLFTPLALPLDYSHYISLPAGVLSLACCTLYGISWQFDPCCKYQVEYDAYKLSRLPLHTLTSSTPVVLVRKDDLHRKRLHNTIALAALVYCVKKIYELYAV
Purification Method Protein G purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Endocrinology, GPCR, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 8834
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.