missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM106B Mouse anti-Human, Clone: 5D1F8, Proteintech
Mouse Monoclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | TMEM106B |
|---|---|
| Clone | 5D1F8 |
| Concentration | 1.48 mg/mL |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16867435
|
Proteintech
60333-1-IG-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16857435
|
Proteintech
60333-1-IG-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Transmembrane protein 106B (TMEM106B) is a single-pass transmembrane protein that is thought to be a novel risk factor for frontotemporal lobar degeneration (FTLD), a group of clinically, pathologically and genetically heterogeneous disorders associated with atrophy in the frontal lobe and temporal lobe of the brain. The actual role of TMEM106B, and that of the closely related protein TMEM106A are still undetermined.Specifications
| TMEM106B | |
| 1.48 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| TMEM106B, transmembrane protein 106B | |
| TMEM106B | |
| IgG1 | |
| Protein G | |
| Antibody |
| 5D1F8 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| Q9NUM4 | |
| 54664 | |
| TMEM106B Fusion Protein Ag21448 | |
| Primary | |
| -20°C | |
| TMEM106B |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title