missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMEM101 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA565246
This item is not returnable.
View return policy
Description
Immunogen sequence: IDGNVAYWHNT RRVEFWNQMK LLGESVGIFG TAVIL Highest antigen sequence identity to the following orthologs - mouse 94%, rat 94%.
May activate NF-kappa-B signaling pathways.
Specifications
| TMEM101 | |
| Polyclonal | |
| Unconjugated | |
| TMEM101 | |
| 2610511E22Rik; Putative NF-kappa-B-activating protein 130; Tmem101; Transmembrane protein 101 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 84336 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96IK0 | |
| TMEM101 | |
| Recombinant Human TMEM101. Recombinant protein control fragment (Product #RP-105967). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction