missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEM1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-48963
This item is not returnable.
View return policy
Description
TMEM1 Polyclonal antibody specifically detects TMEM1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| TMEM1 | |
| Polyclonal | |
| Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| EHOC-1FLJ55683, EHOC1GT334, Epilepsy holoprosencephaly candidate 1 protein, epilepsy holoprosencephaly candidate-1 protein, FLJ54223, FLJ54817, Protein GT334, TMEM1MGC126777, trafficking protein particle complex 10, trafficking protein particle complex subunit 10, trafficking protein particle complex subunit 130, Trafficking protein particle complex subunit TMEM1, transmembrane protein 1, Transport protein particle subunit TMEM1, TRAPP 130 kDa subunit, TRAPP subunit TMEM1, TRS130, TRS30 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: RAVVYSNTREQSSEAALRIQSSDKVTSISLPVAPAYHVIEFELEVLSLPSAPALGGESDMLGMAEPHRKHKDKQRTGRCMVTTDHKVSIDCPWSIYSTVIALTFSVPFRTTHSLLSSGTRKYVQVCVQNLS | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 7109 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction