missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEFF2/Tomoregulin-2 Antibody (Janssen patent anti-TMEFF2), Alexa Fluor™ 405, Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals NBP3-27983AF405
This item is not returnable.
View return policy
Description
TMEFF2/Tomoregulin-2 Monoclonal antibody specifically detects TMEFF2/Tomoregulin-2 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| TMEFF2/Tomoregulin-2 | |
| Monoclonal | |
| Alexa Fluor 405 | |
| CT120.2, HPP1member 2, Hyperplastic polyposis protein 1, TPEFtomoregulin, TR-2, transmembrane protein TENB2, Transmembrane protein with EGF-like and two follistatin-like domains, transmembrane protein with EGF-like and two follistatin-like domains 2, TRtomoregulin-2 | |
| TMEFF2 | |
| 0.1 mL | |
| Cancer, Cell Biology, Cytokines and Growth Factors | |
| 23671 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Janssen patent anti-TMEFF2 | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction