missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMEFF2 Monoclonal Antibody (OTI1E1)
Mouse Monoclonal Antibody
Brand: Invitrogen™ MA525925
This item is not returnable.
View return policy
Description
TMEFF2 Monoclonal Antibody for Western Blot, ICC/IF, Flow
TMEFF2 may be a survival factor for hippocampal and mesencephalic neurons. The shedded form up-regulates cancer cell proliferation, probably by promoting ERK1/2 phosphorylation.
Specifications
| TMEFF2 | |
| Monoclonal | |
| 1.25 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9UIK5 | |
| TMEFF2 | |
| Full length human recombinant protein of TMEFF2 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| OTI1E1 | |
| Unconjugated | |
| TMEFF2 | |
| 4832418D20Rik; 7630402F16Rik; cancer/testis antigen family 120, member 2; CT120.2; HPP1; Hyperplastic polyposis protein 1; MNCb-1026; TENB2; TMEFF2; tomoregulin; tomoregulin-2; TPEF; TR; TR-2; transmembrane protein TENB2; transmembrane protein with EGF like and two follistatin like domains 2; transmembrane protein with EGF-like and two follistatin-like domains; transmembrane protein with EGF-like and two follistatin-like domains 2; UNQ178/PRO204 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 23671 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction