missing translation for 'onlineSavingsMsg'
Learn More

TMEFF1/Tomoregulin-1 Antibody (MM0571-7S12), mFluor Violet 450 SE, Novus Biologicals™

Product Code. 18430918 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18430918 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18430918 Supplier Novus Biologicals Supplier No. NBP211926MFV450

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

TMEFF1/Tomoregulin-1 Monoclonal antibody specifically detects TMEFF1/Tomoregulin-1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen TMEFF1/Tomoregulin-1
Applications Western Blot
Classification Monoclonal
Clone MM0571-7S12
Conjugate mFluor Violet 450 SE
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias C9orf2, H7365member 1, tomoregulin-1, TR-1, Transmembrane protein with EGF-like and one follistatin-like domain, transmembrane protein with EGF-like and two follistatin-like domains 1
Host Species Mouse
Immunogen Human recombinant TMEFF1/Tomoregulin-1
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 8577
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.