missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMEFF1/Tomoregulin-1 Antibody (Bluefin patent anti-TMEFF1) [Janelia Fluor™ 635], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28556JF635
This item is not returnable.
View return policy
Description
TMEFF1/Tomoregulin-1 Monoclonal antibody specifically detects TMEFF1/Tomoregulin-1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| TMEFF1/Tomoregulin-1 | |
| Monoclonal | |
| Janelia Fluor 635 | |
| C9orf2, H7365member 1, tomoregulin-1, TR-1, Transmembrane protein with EGF-like and one follistatin-like domain, transmembrane protein with EGF-like and two follistatin-like domains 1 | |
| TMEFF1/ Tomoregulin-1 | |
| 0.1 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 8577 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Bluefin patent anti-TMEFF1 | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction