Learn More
Invitrogen™ TMED9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110495
Description
Immunogen sequence: MAVELGVLLV RPRPGTGLGR VMRTLLLVLW LATRGSALYF HIGETEKKCF IEEIPDETMV IGNYRTQLYD KQREEYQPAT PGLGMFVEVK DPEDKVILAR QYGSEGRFTF TSHTPGEHQI CLHSNSTKFS LFAGGMLRVH LDIQVGEHAN DYAEIAAKDK LSELQLRVRQ LVEQVEQIQK EQNYQRWREE RFRQTSESTN QRVLWWSILQ TLILVAIGVW QMRH.
TMED9 (also named as GMP25, GP25L2 or p25) is a member of the EMP24/GP25L family. TMED9 probably forms hetero-oligomeric complexes with TMED7 (p27), TMED2 (p24), and TMED10 (p23). Located in the endoplasmic reticulum and the Golgi, TMED9 appears to be involved in vesicular protein trafficking, mainly in the early secretory pathway.
Specifications
| TMED9 | |
| Polyclonal | |
| Unconjugated | |
| TMED9 | |
| 2400003B06Rik; glycoprotein 25L2; GMP25; Gp25l2; gp25L2 protein; HSGP25L2G; p24 family protein alpha-2; p24a2; p24alpha2; p25; RGD1307627; TMED9; Transmembrane emp24 domain-containing protein 9; transmembrane emp24 protein transport domain containing 9; transmembrane p24 trafficking protein 9 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 361207, 54732, 67511 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.9 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q5I0E7, Q99KF1, Q9BVK6 | |
| TMED9 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-224 of human TMED9 (NP_059980.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.