missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMED8 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-85934-0.1ML
This item is not returnable.
View return policy
Description
TMED8 Polyclonal specifically detects TMED8 in Human samples. It is validated for Western Blot.
Specifications
| TMED8 | |
| Polyclonal | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 283578 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| FAM15B, family with sequence similarity 15, member B, MGC126559, protein TMED8, transmembrane emp24 domain containing 8, transmembrane emp24 protein transport domain containing 8 | |
| The immunogen is a synthetic peptide directed towards the middle region of human TMED8 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Vous avez repéré une opportunité d'amélioration ?Partager une correction de contenu