missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMED3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567440
This item is not returnable.
View return policy
Description
Immunogen sequence: DPQGNTIYRET KKQYDSFTYR AEVKGVYQF Highest antigen sequence identity to the following orthologs - mouse 80%, rat 83%.
Potential role in vesicular protein trafficking, mainly in the early secretory pathway. Contributes to the coupled localization of TMED2 and TMED10 in the cis-Golgi network. [UniProt].
Specifications
| TMED3 | |
| Polyclonal | |
| Unconjugated | |
| TMED3 | |
| 1200002G13Rik; AW546672; C15orf22; integral type I protein; membrane protein p24B; MGC133022; p24 family protein gamma-4; P24b; p24g4; p24gamma4; p26; testis tissue sperm-binding protein Li 48e; TMED3; transmembrane emp24 domain containing 3; transmembrane emp24 domain-containing protein 3; transmembrane emp24 domain-containing protein 3-like; transmembrane emp24 protein transport domain containing 3; transmembrane p24 trafficking protein 3; UNQ5357/PRO1078 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 23423 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9Y3Q3 | |
| TMED3 | |
| Recombinant Human TMED3. Recombinant protein control fragment (Product #RP-105602). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction