missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMCO1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA543350
This item is not returnable.
View return policy
Description
Peptide sequence: CSFIFLYILC TMSIRQNIQK ILGLAPSRAA TKQAGGFLGP PPPSGKFS Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 79%.
TMCO1 belongs to the TMCO1 family. The exact function of TMCO1 remains unknown.
Specifications
| TMCO1 | |
| Polyclonal | |
| Unconjugated | |
| TMCO1 | |
| 1190006A08Rik; 4930403O06Rik; AA109065; AU022572; calcium load-activated calcium channel; CLAC channel; ESTM39; HP10122; hypothetical protein LOC614715; LOW QUALITY PROTEIN: transmembrane and coiled-coil domain-containing protein 1; meg-2-like protein; PCIA3; PNAS-10; PNAS-136; putative membrane protein; sb:cb729; TMCC4; tmco1; transmembrane and coiled-coil domain-containing 1-like protein; transmembrane and coiled-coil domain-containing protein 1; transmembrane and coiled-coil domains 1; transmembrane and coiled-coil domains 4; transmembrane and coiled-coil domains protein 1; transmembrane and coiled-coil domains protein 1-like; Transmembrane and coiled-coil domains protein 4; UNQ151/PRO177; Xenogeneic cross-immune protein PCIA3; zgc:110322; zgc:110322 protein; zgc:92740 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 54499 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9UM00 | |
| TMCO1 | |
| Synthetic peptide directed towards the C-terminal of human TMCO1 (aa 192-239). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction