missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TMCC2 Polyclonal antibody specifically detects TMCC2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | TMCC2 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Immunohistochemistry-Paraffin 1:50-1:200 |
| Formulation | PBS (pH 7.0) |
| Gene Alias | cerebral protein 11, FLJ38497, hucep-11, HUCEP11, KIAA0481, transmembrane and coiled-coil domain family 2, transmembrane and coiled-coil domains 2, transmembrane and coiled-coil domains protein 2 |
| Host Species | Rabbit |
| Immunogen | Produced in rabbits immunized with E. coli-derived Human TMCC2 fragment. |
| Purification Method | Antigen and protein A Affinity-purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?