missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TMCC1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 21099-1-AP-20UL
This item is not returnable.
View return policy
Description
TMCC1 is a multi-pass membrane protein. It belongs to the TEX28 family. The exact function of TMCC1 remains unknown.Specifications
| TMCC1 | |
| Polyclonal | |
| Unconjugated | |
| TMCC1 | |
| KIAA0779, TMCC1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 23023, 312654, 330401 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O94876, Q69ZZ6 | |
| Tmcc1 | |
| TMCC1 Fusion Protein Ag15367 | |
| 20 μL | |
| Primary | |
| Mouse, Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction