missing translation for 'onlineSavingsMsg'
Learn More

TMC647055 Choline salt, MedChemExpress

Product Code. 16402747
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16402747 50 mg 50mg
16486816 100 mg 100mg
16469217 5 mg 5mg
16466658 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16402747 Supplier Medchem Express Supplier No. HY15591A
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress TMC647055 choline salt is a cell-permeating, selective HCV NS5B inhibitor, eliciting a mean IC50 of 34 nM, as assessed in the RdRp primer-dependent transcription assay.

TMC647055 choline salt is a cell-permeating, selective HCV NS5B inhibitor, eliciting a mean IC50 of 34 nM, as assessed in the RdRp primer-dependent transcription assay.

Specifications

Chemical Name or Material TMC647055 Choline salt
Color Off-White
Purity Grade Notes Research
Molecular Formula C37H53N5O8S
Quantity 50 mg
Solubility Information DMSO : ≥ 43 mg/mL (59.07 mM)
SMILES OCC[N+](C)(C)C.COC1=CC2=C(C=C1)C3=C(C4CCCCC4)C5=CC=C(C6=O)C=C5N3CC(C(N(C)CCOCCN(S(N6)(=O)=O)C)=O)=C2.[OH-]
Molecular Weight (g/mol) 727.91
Formula Weight 727.91
Percent Purity 98.06%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.