missing translation for 'onlineSavingsMsg'
Learn More

TMC6 Antibody, Janelia Fluor™ 646, Novus Biologicals™

Product Code. 18406033 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18406033 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18406033 Supplier Novus Biologicals Supplier No. NBP176892JF646

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

TMC6 Polyclonal antibody specifically detects TMC6 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Spezifikation

Antigen TMC6
Applications Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 646
Formulation 50mM Sodium Borate
Gene Alias epidermodysplasia verruciformis 1, Epidermodysplasia verruciformis protein 1, EVER1FLJ17776, EVIN1EV1, expressed in activated T/LAK lymphocytes, LAK-4P, Protein LAK-4, transmembrane channel-like 6, transmembrane channel-like protein 6
Host Species Rabbit
Immunogen Antibody was raised against a 15 amino acid synthetic peptide from near the center of human EVER1. The immunogen is located within amino acids 560 - 610 of EVER1. Amino Acid Squence: SEKKLKRRRKPEFDI
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11322
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Mehr anzeigen Weniger anzeigen
Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.