missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMC5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559308
This item is not returnable.
View return policy
Description
Immunogen sequence: TEGRKIMIRL LHEQIINEGK DKMFLIEKLI KLQDMEKKAN PSSLVLERRE VEQQGFLHLG EHDGSLDLRS RRSVQEGNPR Highest antigen sequence identity to the following orthologs: Mouse - 76%, Rat - 78%.
May function as ion channels, transporters, or modulators of such. Ubiquitously expressed.
Specifications
| TMC5 | |
| Polyclonal | |
| Unconjugated | |
| TMC5 | |
| 4932443L08Rik; AU016085; Tmc5; transmembrane channel like 5; transmembrane channel-like 5; transmembrane channel-like gene family 5; transmembrane channel-like protein 5; UNQ8238/PRO33604 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79838 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6UXY8 | |
| TMC5 | |
| Recombinant protein corresponding to Human TMC5. Recombinant protein control fragment (Product #RP-98636). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction