missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TMBIM4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567464
This item is not returnable.
View return policy
Description
Immunogen sequence: RYPRSSIEDDF NYGSSVASAT VHIRMVLQRD CTFSKQYMRI PSAPSATLNI IRFFHLRQSG RSGMVNIFYK GPTFL Highest antigen sequence identity to the following orthologs - mouse 32%, rat 33%.
TMBIM4 is an anti-apoptotic protein which can inhibit apoptosis induced by intrinsic and extrinsic apoptotic stimuli. It can modulate both capacitative Ca2+ entry and inositol 1, 4, 5-trisphosphate (IP3)-mediated Ca2+ release.
Specifications
| TMBIM4 | |
| Polyclonal | |
| Unconjugated | |
| TMBIM4 | |
| 0610007H07Rik; AU022431; Cgi119; CGI-119; CGI-119 protein; GAAP; Golgi anti-apoptotic protein; hypothetical protein LOC513242; LFG4; Protein lifeguard 4; Protein S1R; S1R; Tmbim4; transmembrane BAX inhibitor motif containing 4; transmembrane BAX inhibitor motif-containing protein 4; transmembrane BAX inhibitor motif-containing protein 4; protein lifeguard 4; ZPRO; Z-protein | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 51643 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9HC24 | |
| TMBIM4 | |
| Recombinant Human TMBIM4. Recombinant protein control fragment (Product #RP-108119). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction