missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- Species Reactivity: Species independent
- Storage: Store in the dark
Specifications
Specifications
| Product Type | ELISA Substrate |
| Quantity | 120 mL |
| For Use With (Application) | ELISA |
| Content And Storage | Store in the dark |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?