missing translation for 'onlineSavingsMsg'
Learn More

TMB, MedChemExpress

Product Code. 16491579
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16491579 1 g 1g
16489457 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16491579 Supplier Medchem Express Supplier No. HY15930
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress TMB (BM blue) is a chromogenic substrate used in staining procedures in immunohistochemistry as well as being a visualising reagent used in enzyme-linked immunosorbent assays (ELISA).

TMB (BM blue) is a chromogenic substrate used in staining procedures in immunohistochemistry as well as being a visualising reagent used in enzyme-linked immunosorbent assays (ELISA).

Specifications

Chemical Name or Material TMB
CAS 54827-17-7
Purity Grade Notes Research
Molecular Formula C16H20N2
Synonym BM blue Sure Blue TMB
Solubility Information DMSO : 100 mg/mL (416.08 mM; Need ultrasonic)
SMILES NC1=C(C)C=C(C2=CC(C)=C(N)C(C)=C2)C=C1C
Molecular Weight (g/mol) 240.34
Formula Weight 240.34
Percent Purity 99.83%
Grade Research
Color Off-White
Quantity 1 g
Physical Form Solid
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.