missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TM9SF4 Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
| Antigen | TM9SF4 |
|---|---|
| Concentration | 0.29 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16884155
|
Proteintech
25595-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16874155
|
Proteintech
25595-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
TM9SF4 is a protein coding gene.Specifications
| TM9SF4 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human | |
| Q92544 | |
| 9777 | |
| TM9SF4 Fusion Protein Ag22335 | |
| Primary | |
| -20°C | |
| TM9SF4 |
| 0.29 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| KIAA0255, TM9SF4 | |
| TM9SF4 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title