missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TM9SF3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA534267
Questo articolo non è restituibile.
Consulta la politica di reso
Description
TM9SF3 Polyclonal Antibody for IHC (P)
TM9SF3 is a protein coding gene.
Specifications
| TM9SF3 | |
| Polyclonal | |
| Unconjugated | |
| TM9SF3 | |
| 1810073M23Rik; 2810031D16Rik; AI115521; AI413748; AW146116; AW549777; dinucleotide oxidase disulfide thiol exchanger 3 superfamily member 3; endomembrane protein emp70 precursor isolog; EP70-P-iso; fb75h12; hypothetical protein LOC406698; mKIAA4036; P70-P-iso; RGD1564625; RP11-34E5.1; SM-11044 binding protein; SM-11044-binding protein; SMBP; Tm9sf3; Transmembrane 9 superfamily member 3; Transmembrane 9 superfamily member 3-like protein; transmembrane protein 9 superfamily member 3; UNQ245/PRO282; wu:fa94f05; wu:fb75h12; zgc:56246 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 100070803, 100157588, 100344376, 100714083, 107358, 309475, 395460, 56889, 612786, 614147 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 0.1% sodium azide | |
| Q9ET30, Q9HD45 | |
| TM9SF3 | |
| Synthetic 15 amino acid peptide from internal region of human TM9SF3. | |
| 50 μg | |
| Primary | |
| Human, Mouse, Rat, Canine, Rabbit, Monkey, Bovine, Pig, Horse, Chicken, Guinea Pig, Frog | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction