missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: SLPFCVGSKK SISHYHETLG EALQGVELEF SGLDIKFKDD VMPATYCEID LDKEKRDAFV YAIKNHYWYQ MYIDDLPIWG Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 99%.
TM9SF3 is a protein coding gene.
Specifications
Specifications
| Antigen | TM9SF3 |
| Applications | Immunohistochemistry (Paraffin), Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | TM9SF3 |
| Gene Accession No. | Q9HD45 |
| Gene Alias | 1810073M23Rik; 2810031D16Rik; AI115521; AI413748; AW146116; AW549777; dinucleotide oxidase disulfide thiol exchanger 3 superfamily member 3; endomembrane protein emp70 precursor isolog; EP70-P-iso; fb75h12; hypothetical protein LOC406698; mKIAA4036; P70-P-iso; RGD1564625; RP11-34E5.1; SM-11044 binding protein; SM-11044-binding protein; SMBP; Tm9sf3; Transmembrane 9 superfamily member 3; Transmembrane 9 superfamily member 3-like protein; transmembrane protein 9 superfamily member 3; UNQ245/PRO282; wu:fa94f05; wu:fb75h12; zgc:56246 |
| Gene Symbols | TM9SF3 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?