Learn More
Invitrogen™ TM7SF4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA589951
Description
Immunogen sequence: KILVSASFYP SVERKRIQYL HAKLLKKRSK QPLGEVKRRL SLYLTKIHFW LPVLKMIRKK QMDMASADKS; Positive Samples: 293T; Cellular Location: Cell membrane, Endoplasmic reticulum membrane, Endoplasmic reticulum-Golgi intermediate compartment membrane, Endosome, Multi-pass membrane protein.
DCSTAMP gene ontology annotations related to this gene include cellular response to interleukin-4; cellular response to macrophage colony-stimulating factor stimulus; cellular response to tumor necrosis factor; membrane fusion; myeloid dendritic cell differentiation; negative regulation of cell growth; osteoclast fusion; positive regulation of bone resorption; positive regulation of macrophage fusion; positive regulation of monocyte differentiation.
Specifications
| TM7SF4 | |
| Polyclonal | |
| Unconjugated | |
| DCSTAMP | |
| 4833414I07Rik; DC-specific transmembrane protein; Dcstamp; DC-STAMP; Dendritic cells (DC)-specific transmembrane protein; Dendritic cell-specific transmembrane protein; dendrocyte expressed seven transmembrane protein; dendrocyte-expressed seven transmembrane protein; dentrocyte expressed seven transmembrane protein; FIND; hDC-STAMP; IL-4-induced protein; IL-Four (IL-4) induced; IL-Four INDuced; IL-four-induced protein; LOW QUALITY PROTEIN: dendritic cell-specific transmembrane protein; mDC-STAMP; TM7SF4; transmembrane 7 superfamily member 4 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 75766, 81501 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.71 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q7TNJ0, Q9H295 | |
| DCSTAMP | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 401-470 of human DCSTAMP (NP_1104151). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.