missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ TM7SF2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5110492
This item is not returnable.
View return policy
Description
Immunogen sequence: LAPGGNSGNP IYDFFLGREL NPRICFFDFK YFCELRPGLI GWVLINLALL MKEAELRGSP SLAMWLVNGF QLLYVGDALW HEEAVLTTMD I.
TM7SF2 is involved in the conversion of lanosterol to cholesterol.
Specifications
| TM7SF2 | |
| Polyclonal | |
| Unconjugated | |
| TM7SF2 | |
| 3110041O18Rik; 3-beta-hydroxysterol Delta (14)-reductase; ANG1; Another new gene 1 protein; C-14 sterol reductase; C14SR; delta(14)-sterol reductase; Delta(14)-sterol reductase TM7SF2; delta(14)-sterol reductase TM7SF2; delta(14)-sterol reductase; delta-14-SR; Dhcr14; DHCR14A; NET47; Putative sterol reductase SR-1; sterol C14-reductase; Tm7sf2; Transmembrane 7 superfamily member 2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 7108, 73166 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.16 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| O76062, Q71KT5 | |
| TM7SF2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 160-250 of human TM7SF2 (NP_003264.2). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction