missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TM6SF2 Polyclonal specifically detects TM6SF2 in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | TM6SF2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS, 2% Sucrose |
| Gene Alias | KIAA1926, Lpr4, Transmembrane 6 Superfamily Member 2 |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of Human TM6SF2 |
| Purification Method | Immunogen affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?