missing translation for 'onlineSavingsMsg'
Learn More
Learn More
TM5441, MedChemExpress
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
Description
TM5441 is an orally bioavailable inhibitor of plasminogen activator inhibitor-1 (PAI-1), has IC50 values between 13.9 and 51.1 μM and induces intrinsic apoptosis in several human cancer cell lines. TM5441 attenuates Nω-nitro-l-arginine methyl ester-induced cardiac hypertension and vascular senescence.
Specifications
Specifications
| Chemical Name or Material | TM5441 |
| CAS | 1190221-43-2 |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C21H17ClN2O6 |
| Quantity | 25 mg |
| Solubility Information | DMSO : ≥ 300 mg/mL (699.59 mM) |
| SMILES | ClC1=CC=C(NC(COCC(NC2=CC=CC(C3=COC=C3)=C2)=O)=O)C(C(O)=O)=C1 |
| Molecular Weight (g/mol) | 428.82 |
| Formula Weight | 428.82 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction