missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ TM4SF1 Polyclonal Antibody

Product Code. 12849262
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
12849262 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12849262 Supplier Invitrogen™ Supplier No. PA521119

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

A suggested positive control is human lung tissue lysate. PA5-21119 can be used with blocking peptide PEP-1233.

TM4SF1 is a member of the transmembrane 4 superfamily, also known as the tetraspanin family. Most of these members are cell-surface proteins that are characterized by the presence of four hydrophobic domains and mediate signal transduction events that play a role in the regulation of cell development, activation, growth and motility. TM4SF1 is a cell surface antigen and is highly expressed in the vascular endothelium of several human cancers. Knockdown experiments of TM4SF1 prevented filopodia formation, inhibited cell mobility, blocked cytokinesis, and inhibited maturation of VEGF-A-induced angiogenesis, suggesting that TM4SF1 may be an attractive target for anti-angiogenesis therapy.
TRUSTED_SUSTAINABILITY

Specifications

Antigen TM4SF1
Applications ELISA, Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene TM4SF1
Gene Accession No. P30408, Q64302
Gene Alias 12A8 target antigen; H-L6; L6; L6 antigen; M3S1; Membrane component chromosome 3 surface marker 1; membrane component chromosome 3 surface marker 1 homolog; membrane component, chromosome 3, surface marker 1; membrane component, surface marker 1; TAAL6; TM4SF1; transmembrane 4 L six family member 1; transmembrane 4 L6 family member 1; transmembrane 4 superfamily member 1; tumor-associated antigen L6
Gene Symbols TM4SF1
Host Species Rabbit
Immunogen An 18 amino acid peptide near the amino terminus of human TM4SF1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 17112, 295061, 4071
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.