missing translation for 'onlineSavingsMsg'
Learn More

TM4SF1/L6 Antibody [DyLight 488], Novus Biologicals™

Product Code. 30737812 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30737812 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30737812 Supplier Novus Biologicals Supplier No. AF7514K

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

TM4SF1/L6 Polyclonal antibody specifically detects TM4SF1/L6 in Mouse samples. It is validated for Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen TM4SF1/L6
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias L6, M3S1Tumor-associated antigen L6, Membrane component chromosome 3 surface marker 1, membrane component, chromosome 3, surface marker 1, TAAL6H-L6, transmembrane 4 L six family member 1, transmembrane 4 L6 family member 1, transmembrane 4 superfamily member 1
Host Species Sheep
Immunogen E. coli-derived recombinant mouse TM4SF1, Ala116-Thr161, Accession # Q64302
Purification Method Antigen Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 4071
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.