Learn More
Novus Biologicals™ TM4SF1/L6 Antibody (Beth Israel Patent Anti-TM4SF1) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28479-100ug
 View more versions of this product
Product Code. 30046704
  • £257.00 / 100µg
Description
TM4SF1/L6 Monoclonal antibody specifically detects TM4SF1/L6 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
TM4SF1/L6
Monoclonal
Unconjugated
P30408
Human
Protein A purified
RUO
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1
Flow Cytometry, ELISA, Functional Assay
Beth Israel Patent Anti-TM4SF1
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
L6, M3S1Tumor-associated antigen L6, Membrane component chromosome 3 surface marker 1, membrane component, chromosome 3, surface marker 1, TAAL6H-L6, transmembrane 4 L six family member 1, transmembrane 4 L6 family member 1, transmembrane 4 superfamily member 1
TM4SF1
100 μg
Cancer, Cell Cycle and Replication
4071
Human
Purified