missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
TM2D3 Polyclonal specifically detects TM2D3 in Mouse samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | TM2D3 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS, 2% Sucrose |
| Gene Alias | BBP-like protein 2, BLP2Beta-amyloid-binding protein-like protein 2, FLJ22604, TM2 domain containing 3, TM2 domain-containing protein 3 |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the C-terminal region of TM2D3 |
| Purification Method | Immunogen affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?