missing translation for 'onlineSavingsMsg'
Learn More

TM-N1324, MedChemExpress

Product Code. 16446947
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16446947 5 mg 5mg
16434228 10 mg 10mg
16430567 50 mg 50mg
16405176 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16446947 Supplier Medchem Express Supplier No. HY108699
 Restricted Item
Estimated Shipment: 03-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress TM-N1324 is an agonist of G-Protein-Coupled Receptor 39 (GPR39) with EC50s of 9 nM/5 nM in the presence of Zn2+, and 280 nM/180 nM in the absence of Zn2+ for human/murine GPR39.

TM-N1324 is an agonist of G-Protein-Coupled Receptor 39 (GPR39) with EC50s of 9 nM/5 nM in the presence of Zn2+, and 280 nM/180 nM in the absence of Zn2+ for human/murine GPR39.

Specifications

Chemical Name or Material TM-N1324
CAS 1144477-35-9
Color White
Purity Grade Notes Research
Molecular Formula C18H13ClFN7O
Quantity 5 mg
Solubility Information DMSO : 12.22 mg/mL (30.72 mM; Need ultrasonic)
SMILES O=C1CC(C2=CC=C(F)C=C2Cl)C3=C(N(C4=C5N=CNC5=NC=N4)N=C3C)N1
Molecular Weight (g/mol) 397.79
Formula Weight 397.79
Percent Purity 99.88%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.