missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ TM 5275 sodium salt
Description
TM 5275 sodium salt is a plasminogen activator inhibitor-1 (PAI-1) inhibitor. Orally bioavailable.
Specifications
Specifications
| Chemical Name or Material | 5-Chloro-2-[[2-[2-[4-(Diphenylmethyl)-1-piperazinyl]-2-oxoethoxy]acetyl]amino]benzoic acid sodium salt |
| CAS | 1103926-82-4 |
| Quantity | 50 mg |
| Target | Additional Synthetic Protease Inhibitors |
| Molecular Formula | C28H27ClN3NaO5 |
| Formula Weight | 543.97 |
| Purity | >98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?